Anti-NL1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-4212R-A647
: Bioss
- Antibody type:Primary
- Antigen name:Neuroligin 1
- Antigen symbol:NL1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:22871
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:716-766/863
- Purification:Purified by Protein A
- Pk:100 µl
Cell surface protein involved in cell-cell-interactions via its interactions with neurexin family members. Plays a role in synapse function and synaptic signal transmission, and probably mediates its effects by recruiting and clustering other synaptic proteins. May promote the initial formation of synapses, but is not essential for this. In vitro, triggers the de novo formation of presynaptic structures. May be involved in specification of excitatory synapses (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Neuroligin 1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat