Order Entry
Belgium
ContactUsLinkComponent
Anti-IKBKB Rabbit Polyclonal Antibody (Cy5.5®)
Anti-IKBKB Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-3232R-CY5.5
Supplier:  Bioss
Anti-IKBKB Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-3232R-CY5.5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
  • Antigen symbol:
    IKBKB
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    3551
  • Antigen synonyms:
    IKK-beta|IMD15|IKKB|IKK2|NFKBIKB
  • Modification:
    phospho-Tyr199
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    180-230/756
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Serine kinase that plays an essential role in the NF-kappa-B signaling pathway which is activated by multiple stimuli such as inflammatory cytokines, bacterial or viral products, DNA damages or other cellular stresses. Acts as part of the canonical IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B on 2 critical serine residues. These modifications allow polyubiquitination of the inhibitors and subsequent degradation by the proteasome. In turn, free NF-kappa-B is translocated into the nucleus and activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. In addition to the NF-kappa-B inhibitors, phosphorylates several other components of the signaling pathway including NEMO/IKBKG, NF-kappa-B subunits RELA and NFKB1, as well as IKK-related kinases TBK1 and IKBKE. IKK-related kinase phosphorylations may prevent the overproduction of inflammatory mediators since they exert a negative regulation on canonical IKKs. Also phosphorylates other substrates including NCOA3, BCL10 and IRS1. Within the nucleus, acts as an adapter protein for NFKBIA degradation in UV-induced NF-kappa-B activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IKK beta
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat