Specifications
- Antibody type:Primary
- Antigen name:Transforming Growth Factor beta 5/ Transforming Growth Factor beta 1
- Antigen symbol:TGFB5/ TGFB1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Multiple
- Cross adsorption:No
- Form:liquid
- Gene ID:397778
- Antigen synonyms:Transforming growth factor beta 5|TGF-beta-1|TGF beta 5|Transforming growth factor beta-1.TGFB1_XENLA|Transforming Growth Factor 5|TGF-beta-5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Transforming Growth Factor beta 5 (TGF beta 5) is a member of the TGF beta family of growth factors. The TGF beta polypeptides are multifunctional; capable of influencing cell proliferation, differentiation, and other functions in a wide range of cell types. Transformed, as well as nonneoplastic tissues, release transforming growth factors; and essentially all mammalian cells possess a specific TGF receptor. The multi-modal nature of TGF beta is seen in its ability to stimulate or inhibit cellular proliferation. In general, cells of mesenchymal origin appear to be stimulated by TGF beta whereas cells of epithelial or neuroectodermal origin are inhibited by the peptide.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TGF beta 5/ TGF beta 1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species