Anti-CRX Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-3798R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Cone-rod Homeobox
- Antigen symbol:CRX
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1406
- Antigen synonyms:LCA7|OTX3|CRD|CORD2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:60-110/299
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Transcription factor that binds and transactivates the sequence 5'-TAATC[CA]-3' which is found upstream of several photoreceptor-specific genes, including the opsin genes. Acts synergistically with other transcription factors, such as NRL, RORB and RAX, to regulate photoreceptor cell-specific gene transcription. Essential for the maintenance of mammalian photoreceptors.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CRX
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity: