- Antibody type:Primary
- Antigen name:Thyroid Stimulating Hormone
- Antigen symbol:TSH
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:7252
- Antigen synonyms:follicle-stimulating hormone alpha chain|follicle-stimulating hormone alpha subunit|chorionic gonadotrophin subunit alpha|choriogonadotropin alpha chain|follitropin alpha chain|luteinizing hormone alpha chain|glycoprotein hormones alpha chain|lutropin alpha chain|FSH-alpha|thyrotropin alpha chain|thyroid-stimulating hormone alpha chain|LSH-alpha|alpha polypeptide|TSH-alpha|anterior pituitary glycoprotein hormones common subunit alpha|chorionic gonadotropin
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Thyroid stimulating hormone, also known as thyrotropin, is secreted from cells in the anterior pituitary called thyrotrophs, finds its receptors on epithelial cells in the thyroid gland, and stimulates that gland to synthesize and release thyroid hormones. TSH is a glycoprotein hormone composed of two subunits which are non covalently bound to one another. The alpha subunit of TSH is also present in two other pituitary glycoprotein hormones, follicle stimulating hormone and luteinizing hormone, and, in primates, in the placental hormone chorionic gonadotropin. Each of these hormones also has a unique beta subunit, which provides receptor specificity. In other words, TSH is composed of alpha subunit bound to the TSH beta subunit, and TSH associates only with its own receptor. Free alpha and beta subunits have essentially no biological activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TSH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat