Order Entry
Belgium
ContactUsLinkComponent
 
Anti-TSHB Mouse Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-2676M-FITC
 :  Bioss
Anti-TSHB Mouse Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-2676M-FITC
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Thyroid Stimulating Hormone Beta
  • Antigen symbol:
    TSHB
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rabbit,
    Human,
    Rat
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    7252
  • Antigen synonyms:
    Thyrotropin beta subunit|beta precursor|TSHB.|CHNG4|Thyroid stimulating hormone|Thyroid stimulating hormone beta subunit
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Thyroid stimulating hormone, also known as thyrotropin, is secreted from cells in the anterior pituitary called thyrotrophs, finds its receptors on epithelial cells in the thyroid gland, and stimulates that gland to synthesize and release thyroid hormones. TSH is a glycoprotein hormone composed of two subunits which are non covalently bound to one another. The alpha subunit of TSH is also present in two other pituitary glycoprotein hormones, follicle stimulating hormone and luteinizing hormone, and, in primates, in the placental hormone chorionic gonadotropin. Each of these hormones also has a unique beta subunit, which provides receptor specificity. In other words, TSH is composed of alpha subunit bound to the TSH beta subunit, and TSH associates only with its own receptor. Free alpha and beta subunits have essentially no biological activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSHB
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat, Rabbit