Order Entry
Belgium
ContactUsLinkComponent
Anti-CYP1A2 Rabbit Polyclonal Antibody (Cy7®)
Anti-CYP1A2 Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-2589R-CY7
 :  Bioss
Anti-CYP1A2 Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-2589R-CY7
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Cytochrome P450, Family 1, Subfamily A, Polypeptide 2
  • Antigen symbol:
    CYP1A2
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    1544
  • Antigen synonyms:
    BMD|TU15B|RP50|P3-450|P450(PA)|CP12|VMD2|ARB|BEST
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    245-285/513
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Cytochromes P450 are a group of heme-thiolate monooxygenases. In liver microsomes, this enzyme is involved in an NADPH-dependent electron transport pathway. It oxidizes a variety of structurally unrelated compounds, including steroids, fatty acids, and xenobiotics. Most active in catalyzing 2-hydroxylation. Caffeine is metabolized primarily by cytochrome CYP1A2 in the liver through an initial N3-demethylation. Also acts in the metabolism of aflatoxin B1 and acetaminophen. Participates in the bioactivation of carcinogenic aromatic and heterocyclic amines. Catalizes the N-hydroxylation of heterocyclic amines and the O-deethylation of phenacetin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP1A2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity: