Order Entry
Belgium
ContactUsLinkComponent
Anti-LIG4 Rabbit Polyclonal Antibody (Cy7®)
Anti-LIG4 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-1677R-CY7
Supplier:  Bioss
Anti-LIG4 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-1677R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DNA Ligase 4
  • Antigen symbol:
    LIG4
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    3981
  • Antigen synonyms:
    LIG4S
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    60-110/911
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Efficiently joins single-strand breaks in a double-stranded polydeoxynucleotide in an ATP-dependent reaction. Involved in DNA non-homologous end joining (NHEJ) required for double-strand break repair and V(D)J recombination. The LIG4-XRCC4 complex is responsible for the NHEJ ligation step, and XRCC4 enhances the joining activity of LIG4. Binding of the LIG4-XRCC4 complex to DNA ends is dependent on the assembly of the DNA-dependent protein kinase complex DNA-PK to these DNA ends.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LIG4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat