- Antibody type:Primary
- Antigen name:Golgi Reassembly Stacking Protein 2, 55kDa
- Antigen symbol:GORASP2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:26003
- Antigen synonyms:GOLPH6|GRASP55|GRS2|p59
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
The Golgi apparatus is a highly complex organelle comprised of a stack of cisternal membranes on the secretory pathway from the ER to the cell surface. The structure is maintained by an exoskeleton or Golgi matrix constructed from a family of coiled-coil protein, the golgins and other peripheral membrane components such as GRASP55 and GRASP65 (1). GRASP55 (Golgi reassembly stacking protien or p59) is a component of the Golgi stacking machinery. GRASP55 is highly homologous to GRASP65 and contains two PDZ domains. GRASP55 is myristoylated and palmitoylated. Unlike GRASP65, GRASP55 does not have detectable binding with the vesicle docking protein GM130 and is located on the medial-Golgi rather than cis-Golgi. Both GRASP55 and GRASP65 function in the stacking of Golgi Cisternae (2,3). The novel coiled-coil protein golgin 45 interacts with GRASP55 and the GTP form of Rab 2, suggesting that GRASP55 and golgin 45 form a Rab 2 effector complex on medial-Golgi essential for normal protein transport and Golgi structure (4). ERK2 directly phosphorylates GRASP55, which is phosphorylated in mitotic cells, suggesting that mitogen-activated protein kinase kinase (MKK)/ERK pathway phosphorylates the Golgi during mitosis (5).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Golgi phosphoprotein 6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat