Order Entry
Belgium
ContactUsLinkComponent
Anti-CSTF3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anti-CSTF3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-12935R-A555
Supplier:  Bioss
Anti-CSTF3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-12935R-A555
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cleavage Stimulation Factor, 3' pre-RNA, Subunit 3, 77kDa
  • Antigen symbol:
    CSTF3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    1479
  • Antigen synonyms:
    MGC75122|CSTF 77 kDa subunit|MGC117398|cleavage stimulation factor 3' pre-RNA subunit 3 77kDa|cleavage stimulation factor subunit 3 isoform 1|cleavage stimulation factor subunit 3|MGC43001|CSTF-77
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Polyadenylation of mRNA precursors is a two-step reaction that requires multiple protein factors. The first step, endonucleolytic cleavage of polyadenylation substrates, requires CstF (cleavage stimulation factor), a heterotrimer that is composed of three distinct subunits. Heterotrimeric CstF recognizes GU- and U-rich sequences located downstream of the polyadenylation site on RNA. CstF-77 (cleavage stimulation factor, 77 kDa subunit), also known as CstF3, is one of the three subunits comprising CstF. It can exist as a homodimer and functions as the bridge, directly interacting with the other two CstF subunits, namely CstF-64 and CstF-50. CstF-77 is highly conserved among eukaryotes. It contains an Alpha-helical structure with 11 HAT (Half-a-TPR-containing) repeats and is essential for CstF assembly. In addition, CstF-77 is capable of interacting with CPSF1 and FCP1, other factors involved in polyadenylation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSTF3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity: