Specifications
- Antibody type:Primary
- Antigen name:Metallophosphoesterase Domain Containing 2
- Antigen symbol:MPPED2
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:dJ873F21.1|FAM 1B|Fetal brain protein 239|Chromosome 11 open reading frame 8|Metallophosphoesterase domain containing protein 2|Metallophosphoesterase domain containing 2|dJ1024C24.1|FAM1B|C11orf8|MPPD2_HUMAN.|Hs.46638|D11S302E|239FB|MPPED 2|Brain protein 239
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
MPPED2 (Metallophosphoesterase domain-containing protein 2), also known as C11orf8, FAM1B or 239FB, is a 294 amino acid protein. Expressed primarily in fetal brain tissue, MPPED2 is encoded by a gene that maps to chromosome 11. With approximately 135 million base pairs and 1,400 genes, chromosome 11 makes up around 4% of human genomic DNA and is considered a gene and disease association dense chromosome. The chromosome 11 encoded Atm gene is important for regulation of cell cycle arrest and apoptosis following double stranded DNA breaks. Atm mutation leads to the disorder known as ataxia-telangiectasia. The blood disorders Sickle cell anemia and thalassemia are caused by HBB gene mutations, while Wilms' tumors, WAGR syndrome and Denys-Drash syndrome are associated with mutations of the WT1 gene. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are also associated with defects in chromosome 11.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MPPED2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat