Order Entry
Belgium
ContactUsLinkComponent
Anti-RND2 Rabbit Polyclonal Antibody (Cy7®)
Anti-RND2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-11583R-CY7
Supplier:  Bioss
Anti-RND2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-11583R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Rho family GTPase 2
  • Antigen symbol:
    RND2
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    RND2_HUMAN.|RhoN|Rho N|Rho7|Rho family GTPase 2|Rnd 2|Rho-related GTP-binding protein RhoN|ARHN|Rho 7|Rho related GTP-binding protein RhoN|Rho-related GTP-binding protein Rho7|Rnd2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The Ras p21 family of guanine nucleotide proteins has been widely studied in view of its apparent role in signal transduction pathways and high frequency of mutations in human malignancies. It is now clear, however, that the Ras proteins (H-, K- and N-Ras p21) are members of a much larger superfamily of related proteins. Six members of this family, Rap 1A, Rap 1B, Rap 2, R-Ras, Ral A and Ral B, exhibit approximately 50% amino acid homology to Ras. The six mammalian Rho proteins (Rho A, B, C, G, 7 and 8) are approximately 30% homologous to Ras and are expressed in a wide range of cell types. Both Ras p21 and Rho p21, as well as other members of the Ras superfamily, contain a carboxy-terminal CAAX sequence (C, cysteine; A, aliphatic amino acid; X, any amino acid) which in the case of Ras has been shown to be essential for correct localization and function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RND2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat