Order Entry
Belgium
ContactUsLinkComponent
Anti-CPEB3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-CPEB3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-11340R-A488
Supplier:  Bioss
Anti-CPEB3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-11340R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytoplasmic polyadenylation element binding protein 3
  • Antigen symbol:
    CPEB3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The regulated translation of messenger RNA is essential for cell-cycle progression, establishment of the body plan during early development and modulation of key activities in the central nervous system. Cytoplasmic polyadenylation, one mechanism of controlling translation, is driven by cytoplasmic polyadenylation element binding proteins, called CPEBs. CPEB3 (cytoplasmic polyadenylation element binding protein 3) is a 698 amino acid protein that contains two RNA recognition motif (RRM) domains and, like other CPEB proteins, may play a role in the maturation of the central nervous system. CPEB3 exists as multiple alternatively spliced isoforms that are encoded by a gene which maps to human chromosome 10, which houses over 1,200 genes and comprises nearly 4.5% of the human genome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CPEB3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: