- Antibody type:Primary
- Antigen name:Calsyntenin 2
- Antigen symbol:CLSTN2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:alcagamma|CDHR13|ALC-GAMMA|CS2|CSTN2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Members of the calsyntenin protein family are localized to the post-synaptic membrane of exicitatory central nervous system (CNS) synapses. Calsyntenin-2, also known as Alcadein-gamma, is a 955 amino acid protein that localizes to the endoplasmic reticulum, golgi apparatus and plasma membranes. Containing 2 cadherin-like repeats in its N-terminal extracellular region, calsyntenin-2 binds synaptic calcium with its cytoplasmic domain, suggesting a role in the modulation of calcium-mediated postsynaptic signals. Under normal physiological conditions, calsyntenin-2 is protoeolytically processed in an event in which the primary zeta-cleavage generates a short C-terminal transmembrane fragment and a long extracellular N-terminal domain.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CLSTN2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: