Specifications
- Antibody type:Primary
- Antigen symbol:Nfasc186
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Nfasc186 (Neurofascin 186) is one of two alternatively spliced products of the Neurofascin (Nfasc) gene in rat; the other being Nfasc155. The establishment of paranodal axoglial junctions in myelinated nerves where voltage-gated sodium channels are concentrated at the nodes of Ranvier is determined by myelin-forming glia. The two isoforms of Neurofascin, Nfasc186 in neurons and Nfasc155 in glia, are required for the assembly of these specialized domains. These two major Neurofascins play essential roles in assembling the nodal and paranodal domains of myelinated axons and are essential for the transition to saltatory conduction in developing vertebrate nerves. Nfasc186 (Isoform 1) is expressed at the Nodes of Ranvier while the Nfasc155 (isoforms 2 and 3) are expressed in unmyelinated axons.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: Nfasc186
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat