Anti-PIWIL1 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-0665R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Piwi-Like RNA-Mediated Gene Silencing 1
- Antigen symbol:PIWIL1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:416804
- Antigen synonyms:Piwi-like protein 1|MIWI|PIWI|HIWI|CT80.1|PIWI-L1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:550-600/867
- Purification:Purified by Protein A
- Pk:100 µl