Order Entry
Belgium
ContactUsLinkComponent
Anti-NR3C1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-NR3C1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-0252R-A488
Supplier:  Bioss
Anti-NR3C1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-0252R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nuclear Receptor Subfamily 3 Group C Member 1
  • Antigen symbol:
    NR3C1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    2908
  • Antigen synonyms:
    glucocorticoid nuclear receptor variant 1|GRL|nuclear receptor subfamily 3 group C member 1 variant NS-1|glucocorticoid receptor|GCCR|GCR|GR|nuclear receptor subfamily 3 group C member 1 variant DL-1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    230-280/777
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Receptor for glucocorticoids (GC). Has a dual mode of action: as a transcription factor that binds to glucocorticoid response elements (GRE), both for nuclear and mitochondrial DNA, and as a modulator of other transcription factors. Affects inflammatory responses, cellular proliferation and differentiation in target tissues. Could act as a coactivator for STAT5-dependent transcription upon growth hormone (GH) stimulation and could reveal an essential role of hepatic GR in the control of body growth. Involved in chromatin remodeling. May play a negative role in adipogenesis through the regulation of lipolytic and antilipogenic genes expression.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NR3C1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat