Order Entry
Belgium
ContactUsLinkComponent
Anti-CASP9 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-CASP9 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-0050R-FITC
Supplier:  Bioss
Anti-CASP9 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-0050R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Caspase 9
  • Antigen symbol:
    CASP9
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    842
  • Antigen synonyms:
    EC 3.4.22.62|ICE-like apoptotic protease 6,ICE-LAP6,Apoptotic protease Mch-6,Apoptotic protease-activating factor 3|Apaf-3|CASP-9
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    280-330/416
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Involved in the activation cascade of caspases responsible for apoptosis execution. Binding of caspase-9 to Apaf-1 leads to activation of the protease which then cleaves and activates caspase-3. Promotes DNA damage-induced apoptosis in a ABL1/c-Abl-dependent manner. Proteolytically cleaves poly(ADP-ribose) polymerase (PARP). Isoform 2 lacks activity is an dominant-negative inhibitor of caspase-9.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human