Calibration weight sets, class M1
1 - 200 g, 8, With calibration certificate
Catalog # 611-0965
Supplier: ZWIEBEL
Jump to:
Product Details & Documents
These masses are intended for use calibrating of weights of a lower accuracy class and for use with weighing instruments of medium accuracy class III and and ordinary accuracy class IIII.
Certifications: Conform to the requirements of OIML recommendation R111.
Specifications
- Weight set:1 - 200 g
- Type:With calibration certificate
- No. of weights:8