ContactUsLinkComponent
J.T.Baker® BAKERBOND®, Florisil Bulk Packing Material
J.T.Baker® BAKERBOND®, Florisil Bulk Packing Material
Catalog # 7061-00
Supplier:  Avantor
CAS Number:  
J.T.Baker® BAKERBOND®, Florisil Bulk Packing Material
Catalog # 7061-00
Supplier:  Avantor
CAS Number:  
Specifications
  • Pk:
    100 g
  • Description:
    Florisil SPE sorbent for clean-up columns
Specifications
About this item
SPE sorbent for clean-up columns. Suitable for the determination of Hydrocarbon Oil Index according ISO 9377-2 and NEN 5733.