ContactUsLinkComponent
Reversed phase chromatography columns, RSpak DC-613
 :  Resonac
undefined
Reversed phase chromatography columns, RSpak DC-613
 :  Resonac
  • Catalog No:
  • 554-2874
  • 554-2917
  • Description:
  • RSpak DC-G 4A
  • RSpak DC-613
  • Max. pressure:
  • 5,0 MPa (50 bar)
  • pH range:
  • 2 - 13
  • 2 - 13
  • Temperature limit:
  • 40 - 85 °C
  • 40 - 85 °C
  • USP code:
  • L22, L58
  • L22, L58
Frequently Bought Together