ContactUsLinkComponent
Reversed phase chromatography columns, RSpak NN
Reversed phase chromatography columns, RSpak NN
Supplier:  Resonac
Certificates
undefined
Reversed phase chromatography columns, RSpak NN
Supplier:  Resonac
Specifications
  • Catalog No:
  • 554-2923
  • 554-2885
  • Description:
  • RSpak NN-814
  • RSpak NN-G
  • Max. pressure:
  • 2,0 MPa (20 bar)
  • pH range:
  • 2 - 12
  • 2 - 12
  • Temperature limit:
  • 10 - 60 °C
  • 10 - 50 °C