ContactUsLinkComponent
Reversed phase chromatography columns, RSpak DM-614
Reversed phase chromatography columns, RSpak DM-614
Supplier:  Resonac
Certificates
undefined
Reversed phase chromatography columns, RSpak DM-614
Supplier:  Resonac
Specifications
  • Catalog No:
  • 554-2916
  • 554-2873
  • Description:
  • RSpak DM-614
  • RSpak DM-G 4A
  • Max. pressure:
  • 2,0 MPa (20 bar)
  • pH range:
  • 2 - 10
  • 2 - 10
  • Temperature limit:
  • 20 - 60 °C
  • 20 - 60 °C
  • USP code:
  • L39
  • L39