Cation exchange chromatography columns, TSK-GEL® CM STAT
Supplier: TOSOH Bioscience
Jump to:
About this item
TSK-GEL® CM STAT are cross-linked hydrophilic polymer (mono-disperse particles) based columns designed to analyse the profile of charge isomers of proteins. These STAT columns provide fast, high resolution separations at moderate pressures.
- Nonporous columns
- High efficiency separation at short analysis time
- Used for high resolution protein separation
2 Options Available Below
Product Details & Documents
TSK-GEL® CM STAT are cross-linked hydrophilic polymer (mono-disperse particles) based columns designed to analyse the profile of charge isomers of proteins. These STAT columns provide fast, high resolution separations at moderate pressures.
- Nonporous columns
- High efficiency separation at short analysis time
- Used for high resolution protein separation
Documents
Product Family Options
Product Information
- Column FormatPhaseColumn hardwarePore sizeParticle SizeØ int.LengthPacking materialAvailabilityPrice
- TOYO0021966Cation exchange chromatography column, CM-STAT, nonporousCation exchange chromatography column, CM-STAT, nonporousAnalyticalCM-STATStainless steel-7 µm4,60 mm100 mmPolymerLog in for availability1 * 1 ST1 980,00 €
- TOYO0021965Cation exchange chromatography column, CM-STAT, nonporousCation exchange chromatography column, CM-STAT, nonporousAnalyticalCM-STATStainless steel-10 µm3,00 mm35 mmPolymerLog in for availability1 * 1 ST1 320,00 €