ContactUsLinkComponent
J.T.Baker® BAKERBOND®, Reversed Phase Bulk Packing Material for Preperative HPLC Columns, WP Butyl C4
J.T.Baker® BAKERBOND®, Reversed Phase Bulk Packing Material for Preperative HPLC Columns, WP Butyl C4
Supplier:  Avantor
Certificates
undefined
J.T.Baker® BAKERBOND®, Reversed Phase Bulk Packing Material for Preperative HPLC Columns, WP Butyl C4
Supplier:  Avantor
Certificates
Specifications
  • Average particle size:
    15 µm
  • Working pH:
    2 - 8,5