Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

GET buffer

GET buffer (50 mM glucose, 10 mM EDTA, 25 mM tris HCl, pH 8,0), 100 mL
Catalog # ROCKMB-043
GET buffer
Catalog # ROCKMB-043
CAS Number:  
GET buffer

GET buffer (50 mM glucose, 10 mM EDTA, 25 mM tris HCl, pH 8,0), 100 mL

CAS Number:  
Price
179,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

GET buffer (50 mM Glucose, 10 mM EDTA, 25 mM Tris HCl, pH 8,0) is used in effective plasmid miniprep.

This product is a 1X concentrated stock solution. No dilution is required. Meticulously prepared using ultra pure reagents dissolved in pharmaceutical grade deionised water.

This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    GET buffer (50 mM glucose, 10 mM EDTA, 25 mM tris HCl, pH 8,0)

Specifications