ContactUsLinkComponent
Avantor® Allsep Anion, Anion Exchange HPLC Columns
Avantor® Allsep Anion, Anion Exchange HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Avantor® Allsep Anion, Anion Exchange HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Specifications
  • Catalog No:
  • HICH51200
  • HICH51207
  • 38108/S
  • HICH51208
  • HICH51209
  • HICH51213
  • 38108/P
  • 38109/S
  • HICH38109
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • pH range:
  • 2 - 10
  • 2 - 10
  • 2 - 10
  • 2 - 10
  • 2 - 10
  • USP code:
  • L23
  • L23
  • L23
  • L23
  • L23
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)