Orders LinkContactUsLinkComponent
OmniTop Sample Tubes® Assemblies, Pre-Sterilised Single-Use
OmniTop Sample Tubes® Assemblies, Pre-Sterilised Single-Use
OmniTop Sample Tubes® Assemblies, Pre-Sterilised Single-Use
Certificates
OmniTop Sample Tubes® Assemblies, Pre-Sterilised Single-Use
OmniTop Sample Tubes® Assemblies, Pre-Sterilised Single-Use
12 Options Available Below
Product Details & Documents
OmniTop Sample Tubes® are a convenient device that can be used to obtain fluid samples. Each OmniTop tube comes with a pre-attached 0,2 µm vent filter and 18" of tubing (C-Flex®, TYGON®, silicone or PharMed®).
  • Customisable cap system
  • Unique cap design allows for complete customisation
  • Ability to use different ID and OD tubing Ø
  • Wide selection of tubing materials
  • Easily configured with virtually any type of tubing
  • Single-use
  • Cost effective
  • Reduced assembly and installation time
  • Eliminate cleaning validation
  • Flexible manufacturing
  • Design allows quick delivery on small lots
  • Closure system can be used with various glass bottles
  • Available individually packed or configured in manifold
These tubes are available with or without internal dip-tubes to facilitate removal of the fluid inside.
OmniTop Sample Tubes can be aseptically connected to your bioreactor or other device using standard tubing welders, such as the Terumo SCD-IIB. Sample Tubes can also be aseptically sealed and removed (wet or dry) using tubing sealers such as the Genesis SE340 or handheld battery powered version SE640.
Use OmniTop for the transfer of biopharmaceutical products and reagents in a closed system, gas exchange transfer, sampling of biopharmaceutical products and reagents in a closed system, storage of biopharmaceutical products and reagents. For use with bench top, pilot plant, and production bioreactors. Also for use with sterile connecting devices and thermal or RF tubing sealers.
OmniTop Sample Tubes® are available in a wide variety of standard configurations or they can be customised to suit your specific applications.
Packaging: Available individually packaged or configured in manifold
Product Family Options
OmniTop 15 ml sample tubes
Product Information
  • Capacity
    Material
    Availability
    Price
  • PAWB15CTAPPCFDSL00
    OmniTop assembly, C-Flex®, silicone dip tube, Gamma
    OmniTop assembly, C-Flex®, silicone dip tube, Gamma
    15 ml
    PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 1 090,00 €
    1 090,00 €
    1 * 10 ST
       1 090,00 €
  • 734-2990
    OmniTop assembly, C-Flex®, silicone dip tube, Gamma
    OmniTop assembly, C-Flex®, silicone dip tube, Gamma
    15 ml
    PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 85,30 €
    85,30 €
    1 * 10 ST
       85,30 €
  • 734-2986
    OmniTop assembly, C-Flex®, dip tube, seal and trim, Gamma
    OmniTop assembly, C-Flex®, dip tube, seal and trim, Gamma
    15 ml
    PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 795,00 €
    795,00 €
    1 * 10 ST
       795,00 €
  • 734-2985
    OmniTop assembly, C-Flex®, Gamma
    OmniTop assembly, C-Flex®, Gamma
    15 ml
    PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 13,70 €
    13,70 €
    1 * 10 ST
       13,70 €
  • 734-2987
    OmniTop assembly, C-Flex®, seal and trim, Gamma
    OmniTop assembly, C-Flex®, seal and trim, Gamma
    15 ml
    PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 15,90 €
    15,90 €
    1 * 10 ST
       15,90 €
  • 734-2989
    OmniTop assembly, C-Flex®, Gamma
    OmniTop assembly, C-Flex®, Gamma
    15 ml
    PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 581,00 €
    581,00 €
    1 * 10 ST
       581,00 €
  • 734-2991
    OmniTop assembly, C-Flex®, seal and trim, Gamma
    OmniTop assembly, C-Flex®, seal and trim, Gamma
    15 ml
    PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 8,60 €
    8,60 €
    1 * 10 ST
       8,60 €
OmniTop 50 ml sample tubes
Product Information
  • Capacity
    Material
    Availability
    Price
  • 734-3360
    OmniTop assembly, C-Flex®, silicone dip tube, Gamma
    OmniTop assembly, C-Flex®, silicone dip tube, Gamma
    50 ml
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 620,00 €
    2 620,00 €
    1 * 1 ST
       2 620,00 €
  • 734-2995
    OmniTop assembly, C-Flex®, silicone dip tube, Gamma
    OmniTop assembly, C-Flex®, silicone dip tube, Gamma
    50 ml
    PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 817,00 €
    817,00 €
    1 * 10 ST
       817,00 €
  • PAWB50CTA-CF-DSL-S
    OmniTop assembly, C-Flex®, silicone dip tube, Gamma
    OmniTop assembly, C-Flex®, silicone dip tube, Gamma
    50 ml
    PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 542,00 €
    542,00 €
    1 * 10 ST
       542,00 €
  • 734-2993
    OmniTop assembly, C-Flex®, Gamma
    OmniTop assembly, C-Flex®, Gamma
    50 ml
    PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 52,10 €
    52,10 €
    1 * 10 ST
       52,10 €
  • 734-2994
    OmniTop assembly, C-Flex®, seal and trim, Gamma
    OmniTop assembly, C-Flex®, seal and trim, Gamma
    50 ml
    PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 528,00 €
    528,00 €
    1 * 10 ST
       528,00 €