Order Entry
Belgium
ContactUsLinkComponent
BioRich 1 (DMEM/F12 enriched)
BioRich 1 (DMEM/F12 enriched)
Catalog # ICNA091047122
Supplier:  MP Biomedicals
CAS Number:  
BioRich 1 (DMEM/F12 enriched)
Catalog # ICNA091047122
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Media Format:
    Powder
  • Description:
    BioRich 1 (DMEM/F-12 Enriched)
  • Size:
    1×10 ml
  • Storage temperature:
    4 °C
  • Cat. no.:
    ICNA091047122

Specifications

About this item

BioRich 1 is a DMEM/F-12 powdered media blend enriched with trace elements. This permits the transition to 'serum-free' conditions reducing or eliminating the necessity for serum supplementation.

  • Used with ITS Premix, a completely serum independent

  • Elevated growth of hybridoma lines at reduced serum concentrations

BioRich 1 with 10 common anchorage dependent cell lines commonly show increased yields from 8 to 114% when compared to standard media in normal serum-containing systems. BioRich 1 will support comparable growth to standard media (containing 10% serum) at reduced serum concentrations and in some lines will maintain growth at serum concentrations which will not support growth when using standard media.