Laboratory marker
811-0003
: ELKAY
- Sterile:-
- Colour:Black
- Description:Laboratory marker
- Pk:10
This permanent laboratory marker with medium point can be used to write on glass, plastic, metal and porcelain.
- Even for wet and cold surfaces.
This permanent laboratory marker with medium point can be used to write on glass, plastic, metal and porcelain.