Orders LinkContactUsLinkComponent
Sampling spoons, PharmaSpoons
Sampling spoons, PharmaSpoons
Sampling spoons, PharmaSpoons
Sampling spoons, PharmaSpoons
These sampling spoons are made from stainless steel. They are ideal for transferring small amounts of powder.
  • Crevice-free design
    Bright polished finish
    Head is vertical and side angled (lateral) bent
Available in 316L or 304 stainless steel.
Product Family Options
304 Stainless steel
  • Length
    Spoon size
    Capacity
  • 100 mm
    19×36 mm
    2 ml
    1 * 1 ST
    1 * 1 ST - 20,00 €
    20,00 €
    1 * 1 ST
       20,00 €
  • 210 mm
    19×36 mm
    2 ml
    1 * 1 ST
    1 * 1 ST - 53,80 €
    53,80 €
    1 * 1 ST
       53,80 €
  • 210 mm
    32×65 mm
    10 ml
    1 * 1 ST
    1 * 1 ST - 53,50 €
    53,50 €
    1 * 1 ST
       53,50 €
  • 300 mm
    32×65 mm
    10 ml
    1 * 1 ST
    1 * 1 ST - 74,80 €
    74,80 €
    1 * 1 ST
       74,80 €
  • 500 mm
    32×65 mm
    10 ml
    1 * 1 ST
    1 * 1 ST - 79,80 €
    79,80 €
    1 * 1 ST
       79,80 €
316L Stainless steel
  • Length
    Spoon size
    Capacity
  • 300 mm
    35×70 mm
    10 ml
    1 * 1 ST
    1 * 1 ST - 79,00 €
    79,00 €
    1 * 1 ST
       79,00 €
  • 600 mm
    35×70 mm
    10 ml
    1 * 1 ST
    1 * 1 ST - 95,10 €
    95,10 €
    1 * 1 ST
       95,10 €
  • 1000 mm
    35×70 mm
    10 ml
    1 * 1 ST
    1 * 1 ST - 112,00 €
    112,00 €
    1 * 1 ST
       112,00 €