Order Entry
Belgium
ContactUsLinkComponent
Acetonitrile with 0,1% (v/v) acetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Acetonitrile with 0,1% (v/v) acetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Catalog # 1.59004.2500
Supplier:  Merck
CAS Number:  
Acetonitrile with 0,1% (v/v) acetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Catalog # 1.59004.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
Melting Pt:–45.7 °C
Flash Pt:2 °C
ADR:3,
 :M-Clarity™ quality level = MQ100
UN:1648
 :GHS02
 :GHS07
Storage Temperature:Ambient
 :64-19-7