Screw caps, open top, without septa
Supplier: THERMO NATIONAL SCIENTIFIC
Certificates
About this item
These PP open top screw caps can be used with sample storage screw thread vials.
- PTFE lined solid top
- PTFE on film/foam backing offers broad chemical resistance
- Not autoclavable
Frequently Bought Together