ContactUsLinkComponent
Vaseline soft, lubricant for glass joints, technical grade
Vaseline soft, lubricant for glass joints, technical grade
Catalog # APPC211757.1211
Supplier:  PanReac AppliChem
undefined
Vaseline soft, lubricant for glass joints, technical grade
Catalog # APPC211757.1211
Supplier:  PanReac AppliChem
Specifications
  • Description:
    Lubricating vaseline soft for glass joints
  • Seq:
    02
  • Pk:
    1.000 g
Specifications
About this item
It is a purified mixture of semisolid saturated hydrocarbons having the general formula CnH2n+2 and is obtained from petroleum.
  • White petrolatum
  • Insoluble in water
  • White amorphous mass
Certifications: Meets BP and USP standards.