Order Entry
Belgium
ContactUsLinkComponent
Calcium chloride hexahydrate ≥98% for analysis
Calcium chloride hexahydrate ≥98% for analysis
Catalog # APPC121214.0416
Supplier:  PanReac AppliChem
CAS Number:  
Calcium chloride hexahydrate ≥98% for analysis
Catalog # APPC121214.0416
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Appearence:
    White crystals/crystalline powder
  • Solubility:
    Soluble in water, 5,360 g/l at 20 °C
  • Minimum assay (Compl.) :
    98,0%
  • pH of 5% solution :
    4,5 - 8,5
  • Insoluble matter in H₂O and precip. by NH₄OH:
    0,01%
  • Ammonium (NH₄):
    0,005%
  • Phosphate (PO₄):
    0,001%
  • Sulphate (SO₄):
    0,01%
  • Oxidising substances:
    0,003%
  • Heavy metals (as Pb):
    0,0005%
  • Arsenic (As):
    0,0001%
  • Barium (Ba):
    0,005%
  • Copper (Cu):
    0,0005%
  • Iron (Fe):
    0,0025%
  • Magnesium (Mg):
    0,005%
  • Manganese (Mn):
    0,0005%
  • Nickel (Ni):
    0,0005%
  • Lead (Pb):
    0,0005%
  • Strontium (Sr):
    0,01%
  • Zinc (Zn):
    0,001%
  • Storage:
    Store below 20 °C

Specification Test Results

About this item

 :
 :219.08 g/mol
Melting Pt:30 °C
CAS Number:7774-34-7
Formula:CaCl₂·6H₂O
MDL Number:MFCD00149616