Anti-Bcl-7a Rabbit Polyclonal Antibody [clone: N62]
BWRLBS9274
- Antibody type:Primary
- Antigen symbol:Bcl-7a
- Clonality:Polyclonal
- Clone:N62
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 30-84 of Human Bcl-7a.
Bcl-7a (N62) pAb detects endogenous levels of Bcl-7a protein.
Type: Primary
Antigen: Bcl-7a
Clonality: Polyclonal
Clone: N62
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat