Anti-HSD17B13 Rabbit Polyclonal Antibody
Catalog # BWRLBS7998
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:HSD17B13
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human HSD17B13.
HSD17B13 pAb detects endogenous levels of HSD17B13 protein.
Type: Primary
Antigen: HSD17B13
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human