Anti-OSR2 Rabbit Polyclonal Antibody [clone: F118]
BWRLBS9252
- Antibody type:Primary
- Antigen symbol:OSR2
- Clonality:Polyclonal
- Clone:F118
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 80-136 of Human OSR2.
OSR2 (F118) pAb detects endogenous levels of OSR2 protein.
Type: Primary
Antigen: OSR2
Clonality: Polyclonal
Clone: F118
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat