Anti-Na+ CP type IXalpha Rabbit Polyclonal Antibody [clone: R692]
BWRLBS9164
- Antibody type:Primary
- Antigen symbol:Na+ CP type IX alpha
- Clonality:Polyclonal
- Clone:R692
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 660-718 of Human Na+ CP type IXalpha.
Na+ CP type IX? (R692) pAb detects endogenous levels of Na+ CP type Ix? protein.
Type: Primary
Antigen: Na+ CP type IXalpha
Clonality: Polyclonal
Clone: R692
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat