Anti-NDUFS5 Rabbit Polyclonal Antibody [clone: E89]
BWRLBS9154
- Antibody type:Primary
- Antigen symbol:NDUFS5
- Clonality:Polyclonal
- Clone:E89
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 61-110 of Human NDUFS5.
NDUFS5 (E89) pAb detects endogenous levels of NDUFS5 protein.
Type: Primary
Antigen: NDUFS5
Clonality: Polyclonal
Clone: E89
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human