Anti-KIR2.1 Rabbit Polyclonal Antibody [clone: C122]
Catalog # BWRLBS9150
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:KIR2.1
- Clonality:Polyclonal
- Clone:C122
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 90-140 of Human KIR2.1.
KIR2.1 (C122) pAb detects endogenous levels of KIR2.1 protein.
Type: Primary
Antigen: KIR2.1
Clonality: Polyclonal
Clone: C122
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat