Anti-MaxiKbeta2 Rabbit Polyclonal Antibody [clone: I186]
Catalog # BWRLBS9144
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:MaxiK beta2
- Clonality:Polyclonal
- Clone:I186
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 154-203 of Human MaxiKbeta2.
MaxiK?2 (I186) pAb detects endogenous levels of MaxiK?2 protein.
Type: Primary
Antigen: MaxiKbeta2
Clonality: Polyclonal
Clone: I186
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat