Order Entry
Belgium
ContactUsLinkComponent
Anti-Histone H3 Rabbit Polyclonal Antibody [clone: aDi-Methyl R17]
Anti-Histone H3 Rabbit Polyclonal Antibody [clone: aDi-Methyl R17]
Catalog # BWRLBS7247
Anti-Histone H3 Rabbit Polyclonal Antibody [clone: aDi-Methyl R17]
Catalog # BWRLBS7247

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Histone H3
  • Clonality:
    Polyclonal
  • Clone:
    aDi-Methyl R17
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide conjugated to KLH derived from within residues 1-100 of Human Histone H3, aDi-methylated at R17.

Histone H3 (aDi-Methyl R17) pAb detects endogenous levels of histone H3 only when asymmetric Di-Methyl at Arg17.

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone: aDi-Methyl R17
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human; Mouse; Rat; Other (Wide Range)