Anti-Histone H4 Rabbit Polyclonal Antibody [clone: aDi-Methyl R3]
Catalog # BWRLBS7246
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Histone H4
- Clonality:Polyclonal
- Clone:aDi-Methyl R3
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide
Histone H4 (aDi-Methyl R3) pAb detects endogenous levels of histone H4 only when asymmetric Di-Methyl at Arg3.
Type: Primary
Antigen: Histone H4
Clonality: Polyclonal
Clone: aDi-Methyl R3
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human; Mouse; Rat; Other (Wide Range)