Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Mono-Methyl K79]
Catalog # BWRLBS7240
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Histone H3
- Clonality:Polyclonal
- Clone:Mono-Methyl K79
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide conjugated to KLH derived from within residues 1-100 of Human Histone H3, mono methylated at K79.
Histone H3 (Mono-Methyl K79) pAb detects endogenous levels of histone H3 only when mono-methylated at Lys79.
Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone: Mono-Methyl K79
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human; Mouse; Rat; Other (Wide Range)