Order Entry
Belgium
ContactUsLinkComponent
Anti-COL19A1 Rabbit Polyclonal Antibody [clone: Y442]
Anti-COL19A1 Rabbit Polyclonal Antibody [clone: Y442]
Catalog # BWRLBS9133
Anti-COL19A1 Rabbit Polyclonal Antibody [clone: Y442]
Catalog # BWRLBS9133

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    COL19A1
  • Clonality:
    Polyclonal
  • Clone:
    Y442
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 411-464of Human COL19A1.

COL19A1 (Y442) pAb detects endogenous levels of COL19A1 protein.

Type: Primary
Antigen: COL19A1
Clonality: Polyclonal
Clone: Y442
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human