Anti-HC-II Rabbit Polyclonal Antibody [clone: D68]
Catalog # BWRLBS9125
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:HC-II
- Clonality:Polyclonal
- Clone:D68
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 41-90 of Human HC-II.
HC-II (D68) pAb detects endogenous levels of HC-II protein.
Type: Primary
Antigen: HC-II
Clonality: Polyclonal
Clone: D68
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat