Anti-c-Fos Rabbit Polyclonal Antibody [clone: A356]
Catalog # BWRLBS9112
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:c-Fos
- Clonality:Polyclonal
- Clone:A356
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 331-380 of Human c-Fos.
c-Fos (A356) pAb detects endogenous levels of c-Fos protein.
Type: Primary
Antigen: c-Fos
Clonality: Polyclonal
Clone: A356
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat