Anti-p-Smad1 Rabbit Polyclonal Antibody [clone: S187]
Catalog # BWRLBS4873
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Smad1
- Clonality:Polyclonal
- Clone:S187
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Smad1 around the phosphorylation site of Serine 187.
p-Smad1 (S187) pAb detects endogenous levels of Smad1 protein only when phosphorylated at Ser187.
Type: Primary
Antigen: p-Smad1
Clonality: Polyclonal
Clone: S187
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat