Anti-p-c-Rel Rabbit Polyclonal Antibody [clone: S503]
Catalog # BWRLBS4839
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-c-Rel
- Clonality:Polyclonal
- Clone:S503
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human c-Rel around the phosphorylation site of Serine 503.
p-c-Rel (S503) pAb detects endogenous levels of c-Rel protein only when phosphorylated at Ser503.
Type: Primary
Antigen: p-c-Rel
Clonality: Polyclonal
Clone: S503
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human